missing translation for 'onlineSavingsMsg'
Learn More

Acetyl Chloride, TRC

Product Code. 30310713
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30310713 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30310713 Supplier Toronto Research Chemicals Supplier No. TRCA16449625G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Acetyl Chloride
CAS 75-36-5
Molecular Formula C2H3ClO
Quantity 25 g
Synonym Acetic Acid Chloride, Acetic Chloride, Ethanoyl Chloride, AcCl
InChI Formula InChI=1S/C2H3ClO/c1-2(3)4/h1H3
SMILES CC(=O)Cl
IUPAC Name acetyl chloride
Molecular Weight (g/mol) 78.5
Formula Weight 77.9872
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.