missing translation for 'onlineSavingsMsg'
Learn More

Acetyl L-Carnitine Hydrochloride, TRC

Product Code. 30138039
Change view
Click to view available options
Quantity:
2.5 g
10 g
25 g
Unit Size:
10g
2.5g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30138039 10 g 10g
30137113 25 g 25g
30012451 2.5 g 2.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30138039 Supplier Toronto Research Chemicals Supplier No. TRCA17199010G
 Restricted Item
Estimated Shipment: 10-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5080-50-2
Molecular Formula C9 H18 N O4 . Cl
Molecular Weight (g/mol) 239.7
Synonym (2R)-2-(Acetyloxy)-3-carboxy-N,N,N-trimethyl-1-propanaminium Chloride, 2-(Acetyloxy)-3-carboxy-N,N,N-trimethyl-1-propanaminium, chloride, (R)-, Ammonium, (3-carboxy-2-hydroxypropyl)trimethyl-, chloride, acetate, (-)- (8CI), Acetyl L-carnitine hydrochloride, Acetylcarnitine chloride, Branigen, L-Acetylcarnitine chloride, Levacecarnine hydrochloride, Normobren, O-Acetylcarnitine hydrochloride, ST 200, Zibren, l-Acetylcarnitine hydrochloride, 1-Propanaminium, 2-(acetyloxy)-3-carboxy-N,N,N-trimethyl-, chloride (1:1), (2R)-, 1-Propanaminium, 2-(acetyloxy)-3-carboxy-N,N,N-trimethyl-, chloride, (2R)- (9CI), 1-Propanaminium, 2-(acetyloxy)-3-carboxy-N,N,N-trimethyl-, chloride, (R)-
IUPAC Name [(2R)-2-acetyloxy-3-carboxypropyl]-trimethylazanium;chloride
SMILES [Cl-].CC(=O)O[C@H](CC(=O)O)C[N+](C)(C)C

Specifications

Quantity 10 g
InChI Formula InChI=1S/C9H17NO4.ClH/c1-7(11)14-8(5-9(12)13)6-10(2,3)4;/h8H,5-6H2,1-4H3;1H/t8-;/m1./s1
Formula Weight 239.0924
Percent Purity >95%
Chemical Name or Material Acetyl-L-carnitine hydrochloride
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.