missing translation for 'onlineSavingsMsg'
Learn More

Acetyl-beta-methylcholine bromide, 99%

Product Code. 10627172 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10627172 25 g Glass bottle 25g
10368170 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10627172 Supplier Thermo Scientific Acros Supplier No. 226970250
 Restricted Item
Estimated Shipment: 11-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 333-31-3 | C8H18BrNO2 | 240.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 333-31-3
Molecular Formula C8H18BrNO2
Molecular Weight (g/mol) 240.14
MDL Number MFCD00011816
InChI Key MMVPLEUBMWUYIB-UHFFFAOYNA-M
Synonym methacholine bromide, 2-acetoxy-n,n,n-trimethylpropan-1-aminium bromide, acetyl beta-methylcholine bromide, 1-propanaminium, 2-acetyloxy-n,n,n-trimethyl-, bromide, 1-propanaminium, 2-acetyloxy-n,n,n-trimethyl-, bromide 1:1, methacholini bromidum, methacholine bromide nf, c8h18no2.br, mecholyl bromide, methacholinebromide
PubChem CID 92754
IUPAC Name 2-acetyloxypropyl(trimethyl)azanium;bromide
SMILES [Br-].CC(C[N+](C)(C)C)OC(C)=O

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 150°C to 152°C
Color White
Loss on Drying 1% max.
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (on dried substance) (Argentometry)
Packaging Glass bottle
Linear Formula CH3CO2CH(CH3)CH2N(CH3)3Br
Quantity 25 g
Beilstein 04, III, 758
Merck Index 10, 5793
Solubility Information Solubility in water: soluble
Formula Weight 240.14
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material Acetyl-β-methylcholine bromide, 99%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.