missing translation for 'onlineSavingsMsg'
Learn More

Acetylbenzofuran, TRC

Product Code. 30308884
Change view
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30308884 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30308884 Supplier Toronto Research Chemicals Supplier No. TRCA192408500MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Acetylbenzofuran
CAS 1646-26-0
Molecular Formula C10H8O2
Quantity 500 mg
Synonym 1-(1-Benzofuran-2-yl)ethanone, 1-(2-Benzofuryl)-1-ethanone, 1-(Benzofuran-2-yl)ethan-1-one, 2-Acetylbenzo[b]furan, 2-Acetylbenzofuran, 2-Acetylcoumarone, 2-Benzofuranyl Methyl Ketone, 2-Benzofuryl Methyl Ketone, Benzo[b]furan-2-yl Methyl Ketone, NSC 23974, NSC 28904, 1-(2-Benzofuranyl)ethanone
InChI Formula InChI=1S/C10H8O2/c1-7(11)10-6-8-4-2-3-5-9(8)12-10/h2-6H,1H3
SMILES O=C(C=1OC=2C=CC=CC2C1)C
IUPAC Name 1-(benzofuran-2-yl)ethan-1-one
Molecular Weight (g/mol) 160.17
Formula Weight 60.05
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.