missing translation for 'onlineSavingsMsg'
Learn More

Acetylcholine chloride, 98+%

Product Code. 11331537
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11331537 100 g 100g
11321537 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11331537 Supplier Thermo Scientific Alfa Aesar Supplier No. L02168.22
 Restricted Item
Estimated Shipment: 19-11-2026
Add to basket
Edge

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 60-31-1 | C7H16ClNO2 | 181.66 g/mol

Acetylcholine chloride acts as an endogenous neurotransmitter at cholinergic synapses. The compound has been shown to have ophthalmological uses and can be broken down quickly by choliesterases. Acetylcholine influences many basic cells functions, such as mitosis, cells differentiation, cytoskeletal organization, and cell to cell contact, among other functions. It reduces mortality and plasma proinflammatory cytokines in mice with experimentally-induced sepsis.The cholinergicanti-infl ammatory mechanism is probably mediated by interaction of acetylcholine with α7n cholinoreceptor on monocytes, macrophages, and neutrophils, which decreases the levels of proinflammatory cytokines such as TNF-α, IL-1β, and IL-6.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Acetylcholine chloride acts as an endogenous neurotransmitter at cholinergic synapses. The compound has been shown to have ophthalmological uses and can be broken down quickly by choliesterases. Acetylcholine influences many basic cells functions, such as mitosis, cells differentiation, cytoskeletal organization, and cell to cell contact, among other functions. It reduces mortality and plasma proinflammatory cytokines in mice with experimentally-induced sepsis.The cholinergicanti-infl ammatory mechanism is probably mediated by interaction of acetylcholine with α7n cholinoreceptor on monocytes, macrophages, and neutrophils, which decreases the levels of proinflammatory cytokines such as TNF-α, IL-1β, and IL-6.

Solubility
Soluble in water (600 g/L at 20°C), alcohol, DMSO (36 mg/ml at 25°C), and ethanol (36 mg/ml at 25°C). Insoluble in ether.

Notes
Store away from moisture. Store under dry inert gas. Incompatible with heat,oxidizing agents.
TRUSTED_SUSTAINABILITY

Identificadores de productos químicos

CAS 60-31-1
Molecular Formula C7H16ClNO2
Molecular Weight (g/mol) 181.66
MDL Number MFCD00011698
InChI Key JUGOREOARAHOCO-UHFFFAOYSA-M
Synonym acetylcholine chloride, miochol, acecoline, chloroacetylcholine, arterocoline, acecholin, ovisot, ach chloride, azetylcholinchlorid, 2-acetyloxy-n,n,n-trimethylethanaminium chloride
PubChem CID 6060
ChEBI CHEBI:2417
IUPAC Name 2-acetyloxyethyl(trimethyl)azanium;chloride
SMILES CC(=O)OCC[N+](C)(C)C.[Cl-]

Especificaciones

Melting Point 149°C to 152°C
Quantity 100 g
Beilstein 3571875
Sensitivity Very Hygroscopic
Merck Index 14,87
Solubility Information Soluble in water (600g/L at 20°C),alcohol,DMSO (36mg/ml at 25°C),and ethanol (36mg/ml at 25°C). Insoluble in ether.
Formula Weight 181.66
Percent Purity ≥98%
Chemical Name or Material Acetylcholine chloride
compliance-icons
Nombre del producto
  • Acetylcholine chloride
Palabra de advertencia
  • Warning
Clasificación
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Indicaciones de peligro
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Consejos de prudencia
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Otros peligros
  • MIXTURE LIST-Contain: Acetylcholine chloride

RUO – Research Use Only

Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.