missing translation for 'onlineSavingsMsg'
Learn More

Acetylcholine Chloride, 99%

Product Code. 10465365 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
500g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10465365 500 g Glass bottle 500g
10337430 5 g Glass bottle 5g
10553901 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10465365 Supplier Thermo Scientific Acros Supplier No. 159175000
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 60-31-1 | C7H16ClNO2 | 181.66 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 60-31-1
Molecular Formula C7H16ClNO2
Molecular Weight (g/mol) 181.66
MDL Number MFCD00011698
InChI Key JUGOREOARAHOCO-UHFFFAOYSA-M
Synonym acetylcholine chloride, miochol, acecoline, chloroacetylcholine, arterocoline, acecholin, ovisot, ach chloride, azetylcholinchlorid, 2-acetyloxy-n,n,n-trimethylethanaminium chloride
PubChem CID 6060
ChEBI CHEBI:2417
IUPAC Name 2-acetyloxyethyl(trimethyl)azanium;chloride
SMILES CC(=O)OCC[N+](C)(C)C.[Cl-]

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 149.0°C to 152.0°C
Color White
Loss on Drying 1.0% max. (105°C, 3 hrs)
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (on dried basis) (Argentometry)
Packaging Glass bottle
Linear Formula CH3CO2CH2CH2N(CH3)3Cl
Quantity 500 g
Beilstein 04,III,655
Merck Index 15, 82
Solubility Information Solubility in water: soluble in cold water. Other solubilities: decomposes by hot water or alkalies, soluble in alcohol, chloroform (18.2g/L at 23 °C), practically insoluble in ether
Formula Weight 181.66
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material Acetylcholine chloride
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.