missing translation for 'onlineSavingsMsg'
Learn More

Acetylcholine Chloride, TRC

Product Code. 30137211
Change view
Click to view available options
Quantity:
10 mg
50 mg
1 g
Unit Size:
10mg
1g
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30137211 50 mg 50mg
30135296 1 g 1g
17952115 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30137211 Supplier Toronto Research Chemicals Supplier No. TRCA17206050MG
 Restricted Item
Estimated Shipment: 31-07-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 60-31-1
Molecular Formula C7 H16 N O2 . Cl
Molecular Weight (g/mol) 181.66
Synonym Acetylcholine chloride, 2-(Acetyloxy)-N,N,N-trimethylethanaminium chloride, Ethanaminium, 2-(acetyloxy)-N,N,N-trimethyl-, chloride (1:1), Choline acetate (ester), chloride (8CI), Ethanaminium, 2-(acetyloxy)-N,N,N-trimethyl-, chloride (9CI), (2-Hydroxyethyl)trimethylammonium chloride acetate, 2-Acetoxyethyltrimethylammonium chloride, ACh chloride, Acecholin, Acecoline, Acetylcholine chloride, Acetylcholinium chloride, Arterocoline, Miochol, O-Acetylcholine chloride, Ovisot, Trimethyl(2-acetoxyethyl)ammonium chloride
IUPAC Name 2-acetyloxyethyl(trimethyl)azanium;chloride
SMILES [Cl-].CC(=O)OCC[N+](C)(C)C

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C7H16NO2.ClH/c1-7(9)10-6-5-8(2,3)4;/h5-6H2,1-4H3;1H/q+1;/p-1
Formula Weight 181.087
Chemical Name or Material Acetylcholine chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.