missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Acetylcholine-d13 Bromide (N,N,N-trimethyl-d9; 1,1,2,2-d4), CDN
Click to view available options
Quantity:
0.01 g
0.05 g
Unit Size:
0.01g
0.05g
Specifications
Specifications
| Chemical Name or Material | Acetylcholine-D13 (N,N,N-trimethyl-D9, 1,1,2,2-D4) Bromide |
| Molecular Formula | C7 D13 H3 N O2 . Br |
| Quantity | 0.01 g |
| Synonym | 2-Acetyloxy-N,N,N-tri(methyl-d3)ethan-1,1,2,2-d4-aminium bromide (1:1), Acetylcholine-d13 bromide, Acetylcholine-D13 (N,N,N-trimethyl-D9, 1,1,2,2-D4) bromide, Acetylcholine-d13 Bromide (N,N,N-trimethyl-d9, 1,1,2,2-d4) |
| InChI Formula | InChI=1 S/C7H16NO2.BrH/c1-7(9)10-6-5-8(2,3)4;/h5-6H2,1-4H3;1 H/q+1;/p-1/i2D3,3D3,4D3,5D2,6D2; |
| SMILES | [Br-].[2 H]C([2 H])([2 H])[N+](C([2 H])([2 H])[2 H])(C([2 H])([2 H])[2 H])C([2 H])([2 H])C([2 H])([2 H])OC(=O)C |
| IUPAC Name | (2-acetyloxy-1,1,2,2-tetradeuterioethyl)-tris(trideuteriomethyl)azanium;bromide |
| Molecular Weight (g/mol) | 239.1915 |
| Formula Weight | 238.118 g/mol |
| Percent Purity | 99 atom % D, min 98% Chemical Purity |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction