missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Acetylcholinesterase/ACHE Mouse anti-Human, Clone: 872044, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB7574-SP
This item is not returnable.
View return policy
Description
Acetylcholinesterase/ACHE Monoclonal specifically detects Acetylcholinesterase/ACHE in Human samples. It is validated for Immunohistochemistry.Specifications
| Acetylcholinesterase/ACHE | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| acetylcholinesterase, acetylcholinesterase (Yt blood group), AChE, apoptosis-related acetylcholinesterase, ARACHE, EC 3.1.1, EC 3.1.1.7, N-ACHE, YT, Yt blood group | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 43 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
| Immunohistochemistry | |
| 872044 | |
| Immunohistochemistry 8-25 ug/mL | |
| P22303 | |
| ACHE | |
| Chinese hamster ovary cell line CHO-derived recombinant human ACHE Met1-Leu614 Accession # P22303 | |
| 25 μg | |
| Primary | |
| Detects human ACHE in direct ELISAs. | |
| Human | |
| Purified |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction