missing translation for 'onlineSavingsMsg'
Learn More

Acetylpyrazine, 97%, ACROS Organics™

Product Code. 10687542 Shop All Thermo Scientific Chemicals Products
Click to view available options
:
1g; Glass bottle
This item is not returnable. View return policy

Product Code. 10687542

Brand: Thermo Scientific Acros 224660010

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 22047-25-2
CAS Min % 96.0
CAS Max % 100.0
Melting Point 75°C to 78°C
Color Brown-Yellow to Yellow
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Molecular Formula C6H6N2O
MDL Number MFCD00006134
Quantity 1 g
Synonym acetylpyrazine, 2-acetylpyrazine, ethanone, 1-pyrazinyl, 1-pyrazin-2-yl ethan-1-one, methyl pyrazinyl ketone, 2-acetyl pyrazine, ketone, methyl pyrazinyl, 1-pyrazinylethanone, 1-pyrazin-2-yl-ethanone, 1-pyrazin-2-yl ethanone
InChI Key DBZAKQWXICEWNW-UHFFFAOYSA-N
SMILES CC(=O)C1=NC=CN=C1
IUPAC Name 1-pyrazin-2-ylethanone
Molecular Weight (g/mol) 122.127
PubChem CID 30914
Formula Weight 122.13
Percent Purity ≥96.0%
Physical Form Powder
Chemical Name or Material Acetylpyrazine, 97%
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.