missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Acetylpyrazine, TRC
Click to view available options
Quantity:
10 g
Unit Size:
10g
Specifications
Specifications
| Chemical Name or Material | 2-Acetylpyrazine |
| CAS | 22047-25-2 |
| Molecular Formula | C6 H6 N2 O |
| Quantity | 10 g |
| Synonym | Ethanone, 1-(2-pyrazinyl)-, Ethanone, 1-pyrazinyl- (9CI), Ketone, methyl pyrazinyl (6CI,8CI), 1-(2-Pyrazinyl)ethanone, 1-(2-Pyrazinyl)-1-ethanone, 1-(Pyrazin-2-yl)ethanone, 2-Acetyl-1,4-pyrazine, 2-Acetylpyrazine, Acetylpyrazine, Methyl pyrazin-2-yl ketone, NSC 72374, Pyrazine methyl ketone |
| InChI Formula | InChI=1S/C6H6N2O/c1-5(9)6-4-7-2-3-8-6/h2-4H,1H3 |
| SMILES | CC(=O)c1cnccn1 |
| IUPAC Name | 1-pyrazin-2-ylethanone |
| Molecular Weight (g/mol) | 122.1246 |
| Formula Weight | 122.048 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction