missing translation for 'onlineSavingsMsg'
Learn More

Acetylurea, TRC

Product Code. 30007235
Change view
Click to view available options
Quantity:
100 mg
250 mg
1 g
Unit Size:
100mg
1g
250mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30007235 100 mg 100mg
30004568 1 g 1g
30003510 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30007235 Supplier Toronto Research Chemicals Supplier No. TRCA000230100MG
 Restricted Item
Estimated Shipment: 14-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 591-07-1
Molecular Formula C3H6N2O2
Molecular Weight (g/mol) 102.09
Synonym N-(Aminocarbonyl)acetamide, 1-Acetylurea, Acetylcarbamide, Monoacetylurea, N-Acetylurea, NSC 2766
IUPAC Name N-carbamoylacetamide
SMILES CC(=O)NC(=O)N

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C3H6N2O2/c1-2(6)5-3(4)7/h1H3,(H3,4,5,6,7)
Formula Weight 102.0429
Chemical Name or Material Acetylurea
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.