missing translation for 'onlineSavingsMsg'
Learn More

Acetylvanillin, TRC

Product Code. 30315038
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30315038 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30315038 Supplier Toronto Research Chemicals Supplier No. TRCA18978025G
 Restricted Item
Estimated Shipment: 30-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Acetylvanillin
CAS 881-68-5
Molecular Formula C10H10O4
Quantity 25 g
Synonym NSC 25863, NSC 8499, O-Acetylvanillin, 4-(Acetyloxy)-3-methoxybenzaldehyde, 4-Acetoxy-3-methoxybenzaldehyde, 4-Acetoxy-5-methoxybenzaldehyde, 4-Formyl-2-methoxyphenol Acetate, 4-Formyl-2-methoxyphenyl Acetate, 4-O-Acetylvanillin, 4-(Acetyloxy)-3-methoxybenzaldehyde
InChI Formula InChI=1S/C10H10O4/c1-7(12)14-9-4-3-8(6-11)5-10(9)13-2/h3-6H,1-2H3
SMILES COc1cc(C=O)ccc1OC(=O)C
IUPAC Name (4-formyl-2-methoxyphenyl) acetate
Molecular Weight (g/mol) 194.18
Formula Weight 194.0579
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.