missing translation for 'onlineSavingsMsg'
Learn More

Acevaltrate, MedChemExpress

Product Code. 16494927
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16494927 50 mg 50mg
16493936 25 mg 25mg
16491668 5 mg 5mg
16469176 100 mg 100mg
16429258 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16494927 Supplier Medchem Express Supplier No. HYN2070
 Restricted Item
Estimated Shipment: 01-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Acevaltrate inhibits the Na+/K+-ATPase activity in the rat kidney and brain hemispheres with IC50s of 22.8 μM and 42.3 μM, respectively.

Acevaltrate inhibits the Na+/K+-ATPase activity in the rat kidney and brain hemispheres with IC50s of 22.8 μM and 42.3 μM, respectively.

Specifications

Chemical Name or Material Acevaltrate
CAS 25161-41-5
Color White
Purity Grade Notes Research
Molecular Formula C24H32O10
Quantity 50 mg
Solubility Information DMSO : 100 mg/mL (208.12 mM; Need ultrasonic)
SMILES O=C(CC(C)(C)OC(C)=O)O[C@@H]1[C@]2(CO2)[C@]([C@@H]3OC(CC(C)C)=O)([H])C(C(COC(C)=O)=CO3)=C1
Molecular Weight (g/mol) 480.5
Formula Weight 480.5
Percent Purity 99.56%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.