missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Acid Fuchsin, pure, certified

Product Code. 10187223 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
This item is not returnable. View return policy

Product Code. 10187223

Brand: Thermo Scientific Chemicals undefined

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 3244-88-0 | C20H17N3Na2O9S3 | 587.544 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3244-88-0
Molecular Formula C20H17N3Na2O9S3
Molecular Weight (g/mol) 587.544
MDL Number MFCD00013286
InChI Key VCEFFPMSOGDMJX-UHFFFAOYSA-N
Synonym Acid Violet 19, C.I. 42685, Fuchsin Acid, Rubin S
PubChem CID 131852436
IUPAC Name 3-[bis(4-amino-3-sulfophenyl)methylidene]-6-imino-5-methylcyclohexa-1,4-diene-1-sulfonic acid;sodium
SMILES CC1=CC(=C(C2=CC(=C(C=C2)N)S(=O)(=O)O)C3=CC(=C(C=C3)N)S(=O)(=O)O)C=C(C1=N)S(=O)(=O)O.[Na].[Na]

Specifications

Dye Content 55% min.
Merck Index 15, 99
Solubility Information Solubility in water: 143g/L. Other solubilities: slightly soluble in alcohol
Formula Weight 585.52
Grade Pure
Packaging Glass bottle
Color Green
Quantity 100 g
Physical Form Glittering Powder
Chemical Name or Material Acid Fuchsin, Pure
compliance-icons
Product Identifier
  • Acid Fuchsin
Signal Word
  • Warning
Hazard Category
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.