missing translation for 'onlineSavingsMsg'
Learn More

Acid Orange 7 (Technical Grade), TRC

Product Code. 30138638
Change view
Click to view available options
Quantity:
250 mg
500 mg
2.5 g
Unit Size:
2.5g
250mg
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30138638 500 mg 500mg
30138612 2.5 g 2.5g
18080320 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30138638 Supplier Toronto Research Chemicals Supplier No. TRCA305338500MG
 Restricted Item
Estimated Shipment: 19-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 633-96-5
Molecular Formula C16 H11 N2 O4 S . Na
Molecular Weight (g/mol) 350.32
Synonym Orange 2 sodium, Acid Orange (7CI), Acid Orange A (6CI), Benzenesulfonic acid, 4-[(2-hydroxy-1-naphthalenyl)azo]-, monosodium salt (9CI), Benzenesulfonic acid, p-(2-hydroxy-1-naphthylazo)-, sodium salt (6CI), C.I. Acid Orange 7, monosodium salt (8CI), 11550 Orange, 2-Naphthol Orange II, Acid Leather Orange extra, Acid Leather Orange PRW, Acid Orange 7, Acid Orange II, Acilan Orange II, Airedale Orange II, Amacid Orange Y, Amacid Orange Y Conc, AO 7, Apollo Acid Orange II, Atul Acid Orange II, Basacid Orange 280, Basacid Orange 282, Betanaphthol orange, Borunil Orange A 2R, Brasilan Orange A, BTK Orange II, Bucacid Orange A, C Ext. Orange 8, C.I. 15510, C.I. Acid Orange 7, Calcocid Orange Y, Certiqual Orange II, Colacid Orange, Colocid Orange II, Colorosacid Orange, Conacid Orange L, Concorde Acid Orange II, Concorde Leather Orange MT, Covalene Orange II, Curol Orange, D and C Orange 4 Aluminum Lake, D and C Orange No. 4, D&C Orange #4, D&C Orange No. 4, D&C Orange No. 4-307005, D&C Orange No. 4-90121, Daedo Acid Orange 2G, Derma Fur Orange R 125, Dermacid Orange 2R, Diacid Orange II, Dinacid Orange II, Duasyn Acid Orange P, Dyacid Orange II, Dycosacid Orange Yellow II, Dynacid Orange Y, Egacid Orange II, Eniacid Orange II, Erio Orange II, Everacid Orange II, Fenazo Orange, Golden orange II, Hidacid Orange II, Hispacid Orange AF, Ichoacid Orange II, Indacid Orange II, Japan Orange 205, Japan Orange No. 205, Java Orange II, Kemacid Orange II, Keyacid Orange II, Kiton Orange II, Kromon Lake Orange Toner, Lake Orange A, Lake Orange II YS, Leather Orange Extra, Libacid Orange LII, Lurazol Orange E, Lurazol Orange EBR, Lutetia Orange 3JR, Mandarin G, Multacid Orange II, Multicuer Orange II, Naphthalene Lake Orange G, Naphthalene Orange G, Naphthol Orange, Naphthol Red J, Naphtocard Orange II, Neelicol Orange II, Neklacid Orange II, No. 177 Orange Lake, Nubilon Orange R, Orange 2, Orange 2 sodium salt, Orange 205, Orange Extra N, Orange Extra P, Orange II, Orange II 307004, Orange II 90121, Orange II for Lakes, Orange II Special For Lacquer, Orange IIC, Orange IIP, Orange IIS, Orange IISM, Orange No. 205, Orange Toner GRT, Orange Y, Orange YA, Orange Yellow II, Orange YZ, p-(2-Hydroxy-1-naphthylazo)benzenesulfonic acid sodium salt, Pacid Orange II, Peeracid Orange II, Persian Orange, Persian Orange Lake, Persian Orange X, Pure Orange II S, Rifa Acid Orange 2G, Rifa Acid Orange II, Rifa Leather Orange BR, Ritacid Orange II, Romexal Orange, Rybacel Orange A, Sandal Acid Orange II, Sanyo Gum Orange A, Sodium 4-(2-hydroxy-1-naphthylazo)benzenesulfonate, Solar Orange, Special Orange GR, Special Orange H, Sulfacide Orange JR, Symuler Orange Lake 43, Symulon Acid Orange II, Takaoka Acid Orange II, Tangarine Lake X 917, Tertracid Orange II, Triacid Orange II, Tropaeolin OOO, Tropaeolin OOO 2, Tropeolin OOO, Vibra Color Orange AOR 7, Vicoacid Orange II, Vondacid Orange II, Vopsider Orange A, Water Orange 060506, Wool Orange A, β-Naphthol Orange, β-Naphthyl orange, Benzenesulfonic acid, 4-[2-(2-hydroxy-1-naphthalenyl)diazenyl]-, sodium salt (1:1) (ACI)
IUPAC Name sodium;4-[(E)-(2-hydroxynaphthalen-1-yl)diazenyl]benzenesulfonate
SMILES [Na+].Oc1ccc2ccccc2c1N=Nc3ccc(cc3)S(=O)(=O)[O-]

Specifications

Quantity 500 mg
InChI Formula InChI=1S/C16H12N2O4S.Na/c19-15-10-5-11-3-1-2-4-14(11)16(15)18-17-12-6-8-13(9-7-12)23(20,21)22;/h1-10,19H,(H,20,21,22);/q;+1/p-1
Formula Weight 350.0337
Chemical Name or Material Orange II sodium salt
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.