missing translation for 'onlineSavingsMsg'
Learn More

Acid Red 91, TRC

Product Code. 30329136
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30329136 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30329136 Supplier Toronto Research Chemicals Supplier No. TRCA79529810G
 Restricted Item
Estimated Shipment: 09-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Acid Red 91
CAS 548-24-3
Molecular Formula C20H6Br2N2Na2O9
Quantity 10 g
Synonym 4',5'-dibromo-3',6'-dihydroxy-2',7'-dinitro-Spiro[isobenzofuran-1(3H),9'-[9H]xanthen]-3-one sodium salt, 4’,5’-dibromo-2',7'-dinitro-Fluorescein disodium salt, 4’,5’-dibromo-3',6'-dihydroxy-2',7'-dinitro-Spiro[isobenzofuran-1(3H),9'-[9H]xanthen]-3-one disodium salt, C.I. 45400, C.I. Acid Red 91, Dibromodinitrofluorescein sodium, Eosin B, Eosin BA, Eosin scarlet, Eosine B, Eosine BA, Eosine BN, Eosine BNX, Eosine I Bluish, Eosine Scarlet, Saffrosine, Simacid Nitro Eosin 24669
SMILES [Na+].[Na+].[O-]C(=O)c1ccccc1C2=C3C=C(C(=O)C(=C3Oc4c(Br)c([O-])c(cc24)[N+](=O)[O-])Br)[N+](=O)[O-]
Molecular Weight (g/mol) 624.06
Formula Weight 621.8235
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.