missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Acid Red 91, TRC
Specifications
Specifications
| Chemical Name or Material | Acid Red 91 |
| CAS | 548-24-3 |
| Molecular Formula | C20H6Br2N2Na2O9 |
| Quantity | 1 g |
| Synonym | 4',5'-dibromo-3',6'-dihydroxy-2',7'-dinitro-Spiro[isobenzofuran-1(3H),9'-[9H]xanthen]-3-one sodium salt, 4’,5’-dibromo-2',7'-dinitro-Fluorescein disodium salt, 4’,5’-dibromo-3',6'-dihydroxy-2',7'-dinitro-Spiro[isobenzofuran-1(3H),9'-[9H]xanthen]-3-one disodium salt, C.I. 45400, C.I. Acid Red 91, Dibromodinitrofluorescein sodium, Eosin B, Eosin BA, Eosin scarlet, Eosine B, Eosine BA, Eosine BN, Eosine BNX, Eosine I Bluish, Eosine Scarlet, Saffrosine, Simacid Nitro Eosin 24669 |
| SMILES | [Na+].[Na+].[O-]C(=O)c1ccccc1C2=C3C=C(C(=O)C(=C3Oc4c(Br)c([O-])c(cc24)[N+](=O)[O-])Br)[N+](=O)[O-] |
| Molecular Weight (g/mol) | 624.06 |
| Formula Weight | 621.8235 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?