missing translation for 'onlineSavingsMsg'
Learn More

Acid Red 91, TRC

Product Code. 30333739
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
Product Code. Quantity unitSize
30333739 5 g 5g
1 options
This item is not returnable. View return policy

Product Code. 30333739

Brand: Toronto Research Chemicals TRCA7952985G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Acid Red 91
CAS 548-24-3
Molecular Formula C20H6Br2N2Na2O9
Quantity 5 g
Synonym 4',5'-dibromo-3',6'-dihydroxy-2',7'-dinitro-Spiro[isobenzofuran-1(3H),9'-[9H]xanthen]-3-one sodium salt, 4’,5’-dibromo-2',7'-dinitro-Fluorescein disodium salt, 4’,5’-dibromo-3',6'-dihydroxy-2',7'-dinitro-Spiro[isobenzofuran-1(3H),9'-[9H]xanthen]-3-one disodium salt, C.I. 45400, C.I. Acid Red 91, Dibromodinitrofluorescein sodium, Eosin B, Eosin BA, Eosin scarlet, Eosine B, Eosine BA, Eosine BN, Eosine BNX, Eosine I Bluish, Eosine Scarlet, Saffrosine, Simacid Nitro Eosin 24669
SMILES [Na+].[Na+].[O-]C(=O)c1ccccc1C2=C3C=C(C(=O)C(=C3Oc4c(Br)c([O-])c(cc24)[N+](=O)[O-])Br)[N+](=O)[O-]
Molecular Weight (g/mol) 624.06
Formula Weight 621.8235
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.