missing translation for 'onlineSavingsMsg'
Learn More

Epredia™ Acid-Resistant Biopsy Bags, Medium

Product Code. 12667946 Shop All Epredia Products
Change view
Click to view available options
Size:
Small
Medium
Large
Unit Size:
Pack of 250
Product Code. Size unitSize
12667946 Medium Pack of 250
12677946 Small Pack of 250
12687946 Large Pack of 250
3 options
This item is not returnable. View return policy

Product Code. 12667946

Brand: Epredia™ 6774050

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Reduce risk of small specimen loss with Epredia™ Acid-Resistant Biopsy Bags.

  • Allow unrestricted fluid movement around tissue
  • Reduce reagent carryover and pressure artifact often found with sponges
  • Remove specimen rapidly with convenient peel-open bag

Dimensions:

  • Small Biopsy Bags measure 1.2 × 1.7″ (31 × 43mm)
  • Medium Biopsy Bags measure 1.8 × 2.6″ (45 × 65mm)
  • Large Biopsy Bags measure 3 × 3.5″ (75 × 85mm)
TRUSTED_SUSTAINABILITY

Specifications

Product Type Biopsy Bag
Material Nylon, Acid-resistant
Color White
Length (Metric) 45 mm
Width (Metric) 65 mm
Length (English) 1.8 in.
Width (English) 2.6 in.
Size Medium
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.