Learn More
Fisherbrand™ Acid Resistant Plastic Immersion Tub
Product Code. 10739542 Shop All Fisherbrand Products
Change view
Click to view available options
For Use With (Equipment):
Transonic Analogous Ultrasonic Unit, T 1040/H
Unit Size:
Each
Product Code. For Use With (Equipment) unitSize
10739542 Transonic Analogous Ultrasonic Unit, T 1040/H Each
1 options
Product Code. 10739542
Only null left
Add to basket
Add to basket
Helps protect tanks when using harsh chemical cleaning solutions. Fisherbrand™ Acid Resistant Plastic Immersion Tubs are for use with T 1040/H ultrasonic cleaners.
  • Includes lid/cover
Specifications
Type Acid Resistant Tub with Cover
For Use With (Equipment) Transonic Analogous Ultrasonic Unit, T 1040/H
Includes Cover
Material Plastic