Learn More
Abcam Acidic Mammalian Chitinase Activity Kit (Fluorometric)
Product Code. 32113690 Shop All Abcam Products
Change view
Click to view available options
Quantity:
100 Tests
Unit Size:
100 tests
Product Code. Quantity Unit Size
32113690 100 Tests 100 tests
1 options
Product Code. 32113690 Supplier Abcam Supplier No. ab273291100Test
 Restricted Item
Estimated Shipment: 24-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Acidic Mammalian Chitinase Activity Kit (Fluorometric) (ab273291) utilizes the ability of AMCase to cleave a synthetic substrate to release the free fluorophore which can be easily quantified (Ex/Em= 320/445 nm) and it uses a specific Lysis Buffer which distinguishes AMCase activity from CHIT1 ac...
Acidic Mammalian Chitinase Activity Kit (Fluorometric) (ab273291) utilizes the ability of AMCase to cleave a synthetic substrate to release the free fluorophore which can be easily quantified (Ex/Em= 320/445 nm) and it uses a specific Lysis Buffer which distinguishes AMCase activity from CHIT1 activity.
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Quantity 100 Tests