missing translation for 'onlineSavingsMsg'
Learn More

Acivicin, 98+%

Product Code. 15425379
Change view
Click to view available options
Quantity:
10 mg
25 mg
Unit Size:
10mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15425379 10 mg 10mg
15435379 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15425379 Supplier Thermo Scientific Alfa Aesar Supplier No. J63105.MA
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 42228-92-2 | C5H7ClN2O3 | 178.572 g/mol

Inhibits glutamine amidotransferases in purine and pyrimidine synthetic pathways. Tumor growth inhibitor

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Inhibits glutamine amidotransferases in purine and pyrimidine synthetic pathways
  • Tumor growth inhibitor
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 42228-92-2
Molecular Formula C5H7ClN2O3
Molecular Weight (g/mol) 178.572
MDL Number MFCD00058450
InChI Key QAWIHIJWNYOLBE-OKKQSCSOSA-N
Synonym acivicin, antibiotic at 125, acivicine, acivicino, acivicinum, unii-o0x60k76i6, acia, alphas,5s-alpha-amino-3-chloro-2-isoxazoline-5-acetic acid, acivicin usan:inn, acivicine inn-french
PubChem CID 294641
ChEBI CHEBI:74545
IUPAC Name (2S)-2-amino-2-[(5S)-3-chloro-4,5-dihydro-1,2-oxazol-5-yl]acetic acid
SMILES C1C(ON=C1Cl)C(C(=O)O)N

Specifications

Melting Point 209°C to 211°C
Color White
Quantity 10 mg
UN Number UN2811
Solubility Information Soluble in water at 10mg/ml with warming
Formula Weight 178.57
Percent Purity ≥98%
Physical Form Solid
Chemical Name or Material Acivicin

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.