missing translation for 'onlineSavingsMsg'
Learn More

Acridine-9-carboxylic acid hydrate, 97%

Product Code. 10320134 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
Unit Size:
1g
25g
5g
Product Code. Quantity Packaging unitSize
10320134 25 g Glass bottle 25g
10230173 1 g Glass bottle 1g
10035720 5 g Glass bottle 5g
3 options
This item is not returnable. View return policy

Product Code. 10320134

Brand: Thermo Scientific Acros 400220250

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 332927-03-4 | C14H9NO2 | 223.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 332927-03-4
Molecular Formula C14H9NO2
Molecular Weight (g/mol) 223.23
MDL Number MFCD00149578
InChI Key IYRYQBAAHMBIFT-UHFFFAOYSA-N
Synonym 9-acridinecarboxylic acid hydrate, acridine-9-carboxylic acid hydrate, acmc-1afs1, c14h9no2.h2o, acridin-9-carboxylic acid hydrate, acridine-9-carboxylic acid hydrate 1:x
PubChem CID 16211687
SMILES OC(=O)C1=C2C=CC=CC2=NC2=CC=CC=C12

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 220.0°C
Color Green-Yellow to Yellow
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Quantity 25 g
Beilstein 22,V,3,413
Solubility Information Solubility in water: soluble. Other solubilities: ethanol,pyridine,acetic acid,acetonitrile
IUPAC Name acridine-9-carboxylate
Formula Weight 241.25
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material Acridine-9-carboxylic acid hydrate
Show More Show Less
compliance-icons
Product Identifier
  • Acridine-9-carboxylic acid hydrate
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.