missing translation for 'onlineSavingsMsg'
Learn More

Acridine, 97%

Product Code. 11381367
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11381367 25 g 25g
11371367 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11381367 Supplier Thermo Scientific Alfa Aesar Supplier No. L01657.14
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 260-94-6 | C13H9N | 179.222 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 260-94-6
Molecular Formula C13H9N
Molecular Weight (g/mol) 179.222
MDL Number MFCD00005025
InChI Key DZBUGLKDJFMEHC-UHFFFAOYSA-N
Synonym 9-azaanthracene, 2,3-benzoquinoline, acrydine, 10-azaanthracene, akridin, dibenzo b,e pyridine, 2,3,5,6-dibenzopyridine, akridin czech, benzo b quinoline
PubChem CID 9215
ChEBI CHEBI:36420
IUPAC Name acridine
SMILES C1=CC=C2C(=C1)C=C3C=CC=CC3=N2

Specifications

Melting Point 107°C to 111°C
Density 1.005
Boiling Point 346°C
Quantity 25 g
UN Number UN2713
Beilstein 120200
Merck Index 14,122
Formula Weight 179.22
Percent Purity ≥98%
Chemical Name or Material Acridine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.