missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ Acridine Orange hydrochloride

Description
Cell and organelle membrane permeable nucleic acid binding dye. Emits green fluorescence when bound to double-stranded DNA and red fluorescence when bound to RNA or single-stranded DNA. Used in cell cycle and apoptosis studies and as a lysosomal dye.
Specifications
Specifications
| CAS | 65-61-2 |
| Quantity | 50 mg |
| Molecular Formula | C17H20ClN3 |
| Formula Weight | 301.81 |
| Physical Form | Solid |
| Color | Orange |
| Purity | >99% |
| Molecular Weight (g/mol) | 301.82 |
| Chemical Name or Material | Acridine Orange hydrochloride |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?