missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Acridine Orange, pure, ca. 55% dye content

Product Code. 10531521 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10531521 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10531521 Supplier Thermo Scientific Chemicals Supplier No. 300910250
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 10127-02-3 | C17H20Cl3N3Zn | 438.10 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Melting Point 165°C
CAS 10127-02-3
Odor Odorless
Molecular Formula C17H20Cl3N3Zn
MDL Number MFCD00081043,MFCD00081043,MFCD00081043
UN Number 3143
Color Index No. 46005
Synonym 3,6-Bis(dimethylamino)acridine zinc chloride double salt, Basic Orange 14
Color Brown/Red
Solubility Information Soluble in water
Quantity 25 g
InChI Key VADJQOXWNSPOQA-UHFFFAOYSA-L
Physical Form Powder
SMILES [H+].[Cl-].[Cl-].[Cl-].[Zn++].CN(C)C1=CC2=NC3=CC(=CC=C3C=C2C=C1)N(C)C
IUPAC Name zinc;hydron;3-N,3-N,6-N,6-N-tetramethylacridine-3,6-diamine;trichloride
Molecular Weight (g/mol) 438.10
PubChem CID 71774334
Formula Weight 369.94
Percent Purity 55%
Concentration or Composition (by Analyte or Components) N,N,N',N'-Tetramethylacridine-3,6-diamine monohydrochloride, compound with zinc dichloride
Chemical Name or Material Acridine Orange
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.