missing translation for 'onlineSavingsMsg'
Learn More

Acridine, TRC

Product Code. 17930965
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17930965 1 g 1g
30137992 5 g 5g
30137601 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17930965 Supplier Toronto Research Chemicals Supplier No. TRCA1909001G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 260-94-6
Formula Weight 179.0735
InChI Formula InChI=1S/C13H9N/c1-3-7-12-10(5-1)9-11-6-2-4-8-13(11)14-12/h1-9H
IUPAC Name acridine
Molecular Formula C13 H9 N
Molecular Weight (g/mol) 179.22
Percent Purity >95%
SMILES c1ccc2nc3ccccc3cc2c1
Synonym Acridine, 10-Azaanthracene, 2,3-Benzoquinoline, 9-Azaanthracene, Benzo[b]quinoline, Dibenzo[b,e]pyridine, NSC 3408
Chemical Name or Material Acridine
Grade HPLC
Show More Show Less

Specifications

Quantity 1 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.