missing translation for 'onlineSavingsMsg'
Learn More

Acridinium C2 NHS Ester (>90%), TRC

Product Code. 30179882
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
Unit Size:
10mg
25mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30179882 25 mg 25mg
30180012 5 mg 5mg
30180291 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30179882 Supplier Toronto Research Chemicals Supplier No. TRCA19092525MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 177332-37-5
Molecular Formula C28 H23 N2 O6 . C F3 O3 S
Molecular Weight (g/mol) 632.56
IUPAC Name [4-[3-(2,5-dioxopyrrolidin-1-yl)oxy-3-oxopropyl]phenyl] 10-methylacridin-10-ium-9-carboxylate;trifluoromethanesulfonate
SMILES C[n+]1c2ccccc2c(C(=O)Oc3ccc(CCC(=O)ON4C(=O)CCC4=O)cc3)c5ccccc15.[O-]S(=O)(=O)C(F)(F)F

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C28H23N2O6.CHF3O3S/c1-29-22-8-4-2-6-20(22)27(21-7-3-5-9-23(21)29)28(34)35-19-13-10-18(11-14-19)12-17-26(33)36-30-24(31)15-16-25(30)32;2-1(3,4)8(5,6)7/h2-11,13-14H,12,15-17H2,1H3;(H,5,6,7)/q+1;/p-1
Formula Weight 632.1076
Chemical Name or Material Acridinium C2 NHS Ester
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.