missing translation for 'onlineSavingsMsg'
Learn More

Acriflavine hydrochloride

Codice prodotto. 10651832 Sfoglia Tutto Thermo Scientific Chemicals Prodotti
Cambia vista
Click to view available options
Quantity:
10 g
25 g
100 g
Packaging:
Glass bottle
Dimensione della confezione:
100g
10g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Quantity Packaging unitSize
10651832 100 g Glass bottle 100g
10535501 10 g Glass bottle 10g
10545501 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Este artículo no se puede devolver. Vea la política de devoluciones
Código de producto. 10651832 Proveedor Thermo Scientific Acros N.º de proveedor 346841000
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

Denna artikel kan inte returneras. Se returpolicy

Acriflavine hydrochloride, C27H28Cl4N6, CAS Number-69235-50-3, 3,6-diamino-10-methylacridine proflavine trihydrochloride chloride, acrifiavine hydrochloride, unii-1s73vw819c, acriflavinehydrochloride mixtureof3,6-diamino-10-methylacridiniumchloridehydrochlorideand3,6-diaminoacridinehydrochloride | CAS: 69235-50-3 | C27H27Cl3N6 | 541.91 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Identificadores de productos químicos

CAS 69235-50-3
Molecular Formula C27H27Cl3N6
Molecular Weight (g/mol) 541.91
MDL Number MFCD00069039
InChI Key MKLTXAHQKDVBLY-UHFFFAOYSA-N
Synonym unii-1s73vw819c, acrifiavine hydrochloride, 3,6-diamino-10-methylacridine proflavine trihydrochloride chloride, acriflavinehydrochloride mixtureof3,6-diamino-10-methylacridiniumchloridehydrochlorideand3,6-diaminoacridinehydrochloride
PubChem CID 53393742
IUPAC Name acridine-3,6-diamine;10-methylacridin-10-ium-3,6-diamine;chloride;trihydrochloride
SMILES Cl.Cl.[Cl-].NC1=CC2=NC3=CC(N)=CC=C3C=C2C=C1.C[N+]1=C2C=C(N)C=CC2=CC2=CC=C(N)C=C12

Especificaciones

Melting Point 260.0°C
Color Brown-Red to Orange-Red
Loss on Drying 7% max. (105°C, 1 hr.)
Infrared Spectrum Authentic
Packaging Glass bottle
Quantity 100 g
Physical Form Powder
Chemical Name or Material Acriflavine hydrochloride
compliance-icons compliance-icons compliance-icons
Nombre del producto
  • Acriflavine hydrochloride
Palabra de advertencia
  • Danger
Clasificación
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Serious eye damage/eye irritation Category 1
Indicaciones de peligro
  • H318-Causes serious eye damage.
  • H411-Toxic to aquatic life with long lasting effects.
  • H302-Harmful if swallowed.
Consejos de prudencia
  • P264-Wash thoroughly after handling.
  • P273-Avoid release to the environment.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.