Learn More
Cytiva Acro™ 37 TF Vent Device with PTFE Membrane
Product Code. 17104321
Change view
Click to view available options
Quantity:
200/Pk.
24/Pk.
Unit Size:
Pack of 200
Pack of 24
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17104321 24/Pk. Pack of 24
17114321 200/Pk. Pack of 200
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 17104321 Supplier Cytiva Supplier No. 4464
Only null left
Add to basket
Edge
Add to basket
Disposable, multipurpose filters for small volume venting, solvent, and gas filtration.
  • PTFE membrane and polypropylene housing have broad chemical compatibility.
  • This product is not approved for use in medical, clinical, surgical or other patient applications.
TRUSTED_SUSTAINABILITY
Specifications
Material (Membrane) PTFE
Pore Size 0.2 μm
Sterility Non-sterile
Filtration Area 7.5 cm2
Quantity 24/Pk.
Diameter (Metric) 37 mm