Learn More
Cytiva (formerly Pall Lab) Acrodisc One™ PSF Syringe Filters with wwPTFE Membrane
Perform sample clean-up and preparation functions, ensuring the purest samples are analyzed in liquid chromatography, mass spectrometry, and ion chromatography applications. They utilize the water wettable PTFE (wwPTFE) membrane, a universal membrane for solvents and aqueous samples.
Brand:  Cytiva (formerly Pall Lab) AP-4914
Additional Details : Weight : 3.12620kg
 View more versions of this product
Product Code. 17184701
  • 1460.00 € / Pack of 200
Estimated Shipment: 31-07-2024
to see stock.
Add to basket
Description
Description
Acrodisc One PSF syringe filters offer the following advantages:
  • Extend UHPLC column life over 100 times compared to an unfiltered sample
  • Protect expensive instrument detectors and columns
  • Contribute minimal extractable material that contaminates critical data
  • Available pre-filters produce lower back pressure and higher flow rates for easier sample filtration costs and the expenses associated with downtime for maintenance and recalibration
Specifications
Specifications
wwPTFE
Non-sterile
<125 μL
200/Cs.
55°C
0.2 μm
3.9 cm2
Polypropylene GxF
25/AutoPack Tubes
25 mm