missing translation for 'onlineSavingsMsg'
Learn More

Acrolein diethyl acetal, 94%

Product Code. 10607124 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
This item is not returnable. View return policy

Product Code. 10607124

Brand: Thermo Scientific Acros undefined

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 3054-95-3 | C7H14O2 | 130.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3054-95-3
Molecular Formula C7H14O2
Molecular Weight (g/mol) 130.19
MDL Number MFCD00009239
InChI Key MCIPQLOKVXSHTD-UHFFFAOYSA-N
Synonym acrolein diethyl acetal, acrolein diethylacetal, acrolein acetal, 3,3-diethoxy-1-propene, 1-propene, 3,3-diethoxy, propenal diethyl acetal, 3,3-diethoxypropene, 1,1-diethoxy-2-propene, acrolein, diethyl acetal, acrylaldehyde diethyl acetal
PubChem CID 62477
IUPAC Name 3,3-diethoxyprop-1-ene
SMILES CCOC(C=C)OCC

Specifications

CAS Min % 92.0
CAS Max % 100.0
Color Colorless
Density 0.8500g/mL
Boiling Point 120.0°C to 125.0°C
Flash Point 15°C
Infrared Spectrum Authentic
Assay Percent Range 92% min. (GC)
Packaging Glass bottle
Linear Formula H2C=CHCH(OC2H5)2
Refractive Index 1.3970 to 1.4030
Quantity 100 g
Beilstein 01, 727
Fieser 09,158
Specific Gravity 0.85
Solubility Information Solubility in water: .. Other solubilities: miscible in ethanol,diethyl ether
Formula Weight 130.19
Percent Purity 94%
Physical Form Liquid
Chemical Name or Material Acrolein diethyl acetal
Show More Show Less
compliance-icons
Product Identifier
  • Acrolein diethyl acetal
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
Hazard Statement
  • H225-Highly flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.