missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Acrolein Diethyl Acetal, TRC
Click to view available options
Quantity:
1 g
Unit Size:
1g
Specifications
Specifications
| Chemical Name or Material | Acrolein Diethyl Acetal |
| CAS | 3054-95-3 |
| Molecular Formula | C7H14O2 |
| Quantity | 1 g |
| Synonym | 1,1-Diethoxy-2-propene, 3,3-Diethoxy-1-propene, 3,3-Diethoxypropene, Acrylaldehyde Diethyl Acetal, Acrylylaldehyde Diethyl Acetal, NSC 60135, Propenal Diethyl Acetal |
| InChI Formula | InChI=1S/C7H14O2/c1-4-7(8-5-2)9-6-3/h4,7H,1,5-6H2,2-3H3 |
| SMILES | CCOC(OCC)C=C |
| IUPAC Name | 3,3-diethoxyprop-1-ene |
| Molecular Weight (g/mol) | 130.18 |
| Formula Weight | 130.0994 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction