missing translation for 'onlineSavingsMsg'
Learn More

Acrolein Diethyl Acetal, TRC

Product Code. 30327528
Click to view available options
Quantity:
5 g
Unit Size:
5g
This item is not returnable. View return policy

Product Code. 30327528

Brand: Toronto Research Chemicals TRCA1912055G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Acrolein Diethyl Acetal
CAS 3054-95-3
Molecular Formula C7H14O2
Quantity 5 g
Synonym 1,1-Diethoxy-2-propene, 3,3-Diethoxy-1-propene, 3,3-Diethoxypropene, Acrylaldehyde Diethyl Acetal, Acrylylaldehyde Diethyl Acetal, NSC 60135, Propenal Diethyl Acetal
InChI Formula InChI=1S/C7H14O2/c1-4-7(8-5-2)9-6-3/h4,7H,1,5-6H2,2-3H3
SMILES CCOC(OCC)C=C
IUPAC Name 3,3-diethoxyprop-1-ene
Molecular Weight (g/mol) 130.18
Formula Weight 130.0994
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.