missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ AcroMetrix™ HIV-1 Controls

Product Code. 15945950 Shop All Thermo Scientific Products
Change view
Click to view available options
Description:
AcroMetrix HIV-1 Mid Control
Unit Size:
6mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Description Unit Size
15945950 AcroMetrix HIV-1 Mid Control 6mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15945950 Supplier Thermo Scientific™ Supplier No. 964002
  • ONLINE EXCLUSIVE PRICE
    675.00 €
    523.20 € / 6mL
    Save 151.80 €
    22% Off
 Restricted Item
Estimated Shipment: 24-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Identify increases in random or systematic error of nucleic acid test procedures for HIV-1 RNA with these controls, which are for In Vitro Diagnostic Use.

Assess the performance of molecular diagnostic test procedures, including the RNA extraction step, for the quantitative and qualitative determination of human immunodeficiency virus type-1 (HIV-1) RNA with the Thermo Scientific™ AcroMetrix™ HIV-1 Controls. These controls can be used for training and proficiency testing of laboratory personnel, for lot-to-lot comparison of reagent test kits and to evaluate and compare intralaboratory and interlaboratory assessments. The controls are for In Vitro Diagnostic Use.

TRUSTED_SUSTAINABILITY

Specifications

Description AcroMetrix HIV-1 Mid Control
Detectable Analytes HIV-1
Type HIV-1 Control
Packaging 5 x 1.2mL
Quantity 5 x 1.2 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.