Learn More
Cytiva AcroPak™ 1000 Capsules with Supor™ Membrane
Product Code. 17114251
Change view
Click to view available options
Pore Size:
0.1/0.1 μm
0.2/0.2 μm
0.8/0.2 μm
0.8/0.45 μm
Unit Size:
Each
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Pore Size Unit Size
17114251 0.8/0.2 μm Each
17134251 0.8/0.45 μm Each
17154251 0.2/0.2 μm Each
17174251 0.1/0.1 μm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 17114251 Supplier Cytiva Supplier No. 12992
Estimated Shipment: 07-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
AcroPak Capsules for buffers, tissue culture media, and other biological fluids.
  • Newer technology recommended for simplified scale-up
  • High filtration area and compact size are ideal for upscale trials
  • Designed for research applications only
  • Gamma-irradiated
TRUSTED_SUSTAINABILITY
Specifications
Material (Membrane) Hydrophilic PES
Pore Size 0.8/0.2 μm
Sterility Sterile
Filtration Area 1000 cm2
Outlet Style Stepped Hose Barb
Quantity 1/Ea.