Learn More
Cytiva AcroPak™ 1500 Capsules with Supor™ EKV Membrane
Description
- Newer technology recommended for simplified scale-up
- High filtration area and compact size are ideal for upscale trials
- Designed for research applications only
- Gamma-irradiated
Specifications
Specifications
| Material (Membrane) | Hydrophilic PES |
| Pore Size | 0.2 μm |
| Sterility | Sterile |
| Filtration Area | 1500 cm2 |
| Outlet Style | 1/2 in. Hose Barb |
| Quantity | 1/Ea. |