Learn More
Cytiva AcroPak™ 1500 Capsules with Supor™ Membrane
Product Code. 17184241
Change view
Click to view available options
Pore Size:
0.2/0.2 μm
0.8/0.2 μm
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Pore Size Unit Size
17184241 0.2/0.2 μm Each
17174241 0.8/0.2 μm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 17184241 Supplier Cytiva Supplier No. 12686
In Stock
Add to basket
Edge
Add to basket
missing translation for 'validMainframeMsg'
AcroPak Capsules for buffers, tissue culture media, and other biological fluids.
  • Newer technology recommended for simplified scale-up
  • High filtration area and compact size are ideal for upscale trials
  • Designed for research applications only
  • Gamma-irradiated
TRUSTED_SUSTAINABILITY
Specifications
Material (Membrane) Hydrophilic PES
Pore Size 0.2/0.2 μm
Sterility Sterile
Filtration Area 1500 cm2
Outlet Style 1/2 in. Hose Barb
Quantity 1/Ea.