Learn More
Cytiva™ AcroPak™ 200 Capsules with Supor™ Membrane
Product Code. 17194241
Change view
Click to view available options
Outlet Style:
1/4 in. MNPT Inlet, Stepped Hose Barb Outlet
Stepped Hose Barb
Unit Size:
Pack of 3
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Outlet Style Unit Size
17194241 Stepped Hose Barb Pack of 3
17134221 1/4 in. MNPT Inlet, Stepped Hose Barb Outlet Pack of 3
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 17194241 Supplier Cytiva™ Supplier No. 12941
In Stock
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
AcroPak capsules designed for research applications only.
  • Efficient processing of liquid volumes up to 20 L
  • Built-in prefilter extends filter life when particulate-laden solutions, such as serum-containing media, are processed
  • For buffers, tissue culture media, and other biological fluids
  • Newer technology recommended for simplified scale-up
  • High filtration area and compact size are ideal for upscale trials
TRUSTED_SUSTAINABILITY
Specifications
Material (Membrane) Hydrophilic PES
Pore Size 0.8/0.2 μm
Sterility Sterile
Filtration Area 200 cm2
Outlet Style Stepped Hose Barb
Quantity 3/Pk.