Learn More
Cytiva AcroPak™ 400 Capsules with Fluorodyne™ II Membrane
Description
- Double layer sterilizing membrane enhances reliability
- Upstream air vent prevents vapor lock
- Manufactured without the use of adhesives to minimize extractables
- Pre-sterilized by gamma irradiation
Specifications
Specifications
| Material (Membrane) | Hydrophilic PVDF |
| Pore Size | 0.2 μm |
| Sterility | Sterile |
| Filtration Area | 400 cm2 |
| No. per Pack | 1/Ea. |
| Outlet Style | Stepped Hose Barb |