missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva (formerly Pall Lab) AcroPak™ 800 Capsules with Supor™ EKV Membrane
AcroPak capsules designed for research applications only.
228.00 €
Specifications
Material (Membrane) | Hydrophilic PES |
---|---|
Pore Size | 0.2 μm |
Sterility | Sterile |
Filtration Area | 750 cm2 |
Quantity | 1/Ea. |
Product Code | Brand | Outlet Style | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Outlet Style | Price | Quantity & Availability | |||||
17114241
|
Cytiva (formerly Pall Lab)
12464 |
1/2 in. Hose Barb Inlet with Filling Bell on Outlet |
228.00 €
Each |
Estimated Shipment: 03-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
17104241
|
Cytiva (formerly Pall Lab)
12463 |
1 to 1.5 in. Sanitary Flange |
228.00 €
Each |
Estimated Shipment: 03-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
- For buffers, tissue culture media, and other biological fluids
- Newer technology recommended for simplified scale-up
- High filtration area and compact size are ideal for upscale trials
Specifications
Hydrophilic PES | |
Sterile | |
1/Ea. |
0.2 μm | |
750 cm2 |