missing translation for 'onlineSavingsMsg'
Learn More

Cytiva AcroPrep™ Advance 96-well Filter Plates for Aqueous Filtration

Product Code. 17194611
Change view
Click to view available options
Pore Size:
0.2 μm
1.0 μm
30 to 40 μm
Volume (Metric):
350 μL
1 mL
2 mL
350 mL
Material (Membrane):
Borosilicate Glass w/o Binder, Supor (PES)
Polypropylene/Polyethylene Non-woven Media
Supor (PES)
Unit Size:
Pack of 10
Pack of 5
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Pore Size Volume (Metric) Material (Membrane) unitSize
17194611 0.2 μm 1 mL Supor (PES) Pack of 5
17114601 0.2 μm 350 μL Supor (PES) Pack of 10
17164631 1.0 μm 2 mL Borosilicate Glass w/o Binder, Supor (PES) Pack of 5
17134601 30 to 40 μm 350 μL Polypropylene/Polyethylene Non-woven Media Pack of 10
17154601 1.0 μm 350 mL Borosilicate Glass w/o Binder, Supor (PES) Pack of 10
17104621 30 to 40 μm 1 mL Polypropylene/Polyethylene Non-woven Media Pack of 5
17134621 1.0 μm 1 mL Borosilicate Glass w/o Binder, Supor (PES) Pack of 5
17154631 30 to 40 μm 2 mL Polypropylene/Polyethylene Non-woven Media Pack of 5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
This item is not returnable. View return policy
Product Code. 17194611 Supplier Cytiva Supplier No. 8119
Only null left
Add to basket
Edge
Add to basket
This item is not returnable. View return policy

Optimized for a variety of applications requiring larger per well volume processing.

  • Plates are constructed of chemically resistant, biologically-inert polypropylene
  • This product is not approved for use in medical, clinical, surgical or other patient applications
TRUSTED_SUSTAINABILITY

Specifications

Material (Membrane) Supor (PES)
Pore Size 0.2 μm
Sterility Non-sterile
Housing Material Polypropylene Housing
Quantity 5/Pk.
Volume (Metric) 1 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.