Learn More
Thermo Scientific Chemicals Agar, Bacteriological grade
Shop All Thermo Scientific Chemicals ProductsDescription
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Microbial media
Specifications
Specifications
| Form | Powder |
| Grade | Bacteriological |
| Product Type | Agar |
| For Use With (Application) | Microbial media |
| Packaging Type | Plastic Bottle |
| Quantity | 5 kg |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.