missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Acrylamide, 98.5%, extra pure

Product Code. 10216170 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
500 g
2.5 kg
10 kg
Packaging:
Plastic Bottle
Plastic Drum
Unit Size:
10kg
2.5kg
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10216170 500 g Plastic Bottle 500g
10062040 2.5 kg Plastic Bottle 2.5kg
11975461 10 kg Plastic Drum 10kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.
Product Code. 10216170 Supplier Thermo Scientific Chemicals Supplier No. 164835000
Black Friday Starts Here
Use promo code 27984
View all promotions
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.

Acrylamide, CAS # 79-06-1, is an organic amide compound used as a monomer in the synthesis of polyacrylamides. | CAS: 79-06-1 | C3H5NO | 71.08 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

  • Acrylamide is an organic amide compound mainly used to as a monomer in the synthesis of polyacrylamide
  • Acrylamide is formed in foods containing carbohydrates in its composition, mainly when these foods are fried, grilled or baked

Applications

  • In laboratory experiments acrylamide participates in chemical reactions as an intermediate in the preparation of N-methylol acrylamide and N-butoxyacrylamide compounds
  • For molecular usage, this compound can be used as a carrier and is involved in the precipitation of small amounts of DNA
  • In molecular experiments it is used in gel electrophoresis, including western blot, and SDS-PAGE, to separate proteins and macromolecules based on their size and charge
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 79-06-1
Molecular Formula C3H5NO
Molecular Weight (g/mol) 71.08
MDL Number MFCD00008032
InChI Key HRPVXLWXLXDGHG-UHFFFAOYSA-N
Synonym Ethylenecarboxamide, 2-Propenamide
PubChem CID 6579
ChEBI CHEBI:28619
IUPAC Name prop-2-enamide
SMILES NC(=O)C=C

Specifications

Physical Form Crystals or Crystalline Powder
Boiling Point 125.0°C (25.0 mmHg)
Melting Point 82.0°C to 86.0°C
Density 1.1220g/mL
Color White
Quantity 500 g
Flash Point 138°C
Infrared Spectrum Authentic
Packaging Plastic Bottle
Assay Percent Range 98.5%
Linear Formula H2C=CHCONH2
Solubility Information Solubility in water: 21.5g/L (30°C). Other solubilities: solubilities in g/100mL solvent (30°C):, methanol 155; ethanol 86.2; acetone 63.1;, ethyl acetate 12.6; chloroform 2.66;, benzene 0.346; heptane 0.0068, soluble in ether
Formula Weight 71.08
Percent Purity 98.5%
Grade Extra Pure
Chemical Name or Material Acrylamide
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Acrylamide
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Skin sensitiser Category 1
  • Germ cell mutagenicity Category 1B
  • Carcinogenicity Category 1B
  • Reproductive toxicity Category 2
  • Specific target organ toxicity Category 1
Hazard Statement
  • H301-Toxic if swallowed.
  • H372-Causes damage to organs.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H315-Causes skin irritation.
  • H317-May cause an allergic skin reaction.
  • H319-Causes serious eye irritation.
  • H340-May cause genetic defects.
  • H350-May cause cancer.
  • H361f-Suspected of damaging fertility.
Precautionary Statement
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/and soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.