Learn More
Acrylamide, 99.9%
Description
Acrylamide is primarily used in research laboratories for electrophoresis, chromatography, electron microscopy and in the sewage and wastewater treatment industries. It is used in the production of polymers mainly in the pulp and paper industry and textile treatment industry. The largest use for acrylamide is as an intermediate in the production of organic chemicals and in the synthesis of polyacrylamides.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | 82°C to 85°C |
| Density | 1.322 g/mL |
| Boiling Point | 125°C (25 mmHg) |
| Flash Point | 138°C (280°F) |
| Assay Percent Range | 99.9% |
| Odor | Odorless |
| Quantity | 100 g |
| UN Number | UN2074 |
| Beilstein | 605349 |
| Sensitivity | Air and light sensitive |
| Show More |
Safety and Handling
- Acrylamide
- Danger
- Acute toxicity Category 3
- Acute toxicity Category 4
- Carcinogenicity Category 1B
- Serious eye damage/eye irritation Category 2
- Germ cell mutagenicity Category 1B
- Reproductive toxicity Category 2
- Skin corrosion/irritation Category 2
- Skin sensitiser Category 1
- Specific target organ toxicity Category 1
- H301-Toxic if swallowed.
- H312-Harmful in contact with skin.
- H315-Causes skin irritation.
- H317-May cause an allergic skin reaction.
- H319-Causes serious eye irritation.
- H332-Harmful if inhaled.
- H340-May cause genetic defects.
- H350-May cause cancer.
- H361f-Suspected of damaging fertility.
- H372-Causes damage to organs.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
- P337+P313-If eye irritation persists: Get medical advice/attention.
- MIXTURE LIST-Contain: Acrylamide
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.