missing translation for 'onlineSavingsMsg'
Learn More

Acrylamide: Bis-Acrylamide 19:1 (Powder/Electrophoresis), Fisher BioReagents

Product Code. 10786644
Change view
Click to view available options
Quantity:
100 g
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10786644 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10786644 Supplier Fisher BioReagents Supplier No. BP1364100
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
This dry powder mix is premixed for 5% cross-linking and is suitable for DNA sequencing and separation of low-molecular-weight proteins.
TRUSTED_SUSTAINABILITY

Specifica

Physical Form Solid
CAS 79-06-1, 110-26-9
Conductivity 10μmhos max.
Color White
Quantity 100 g
DNase DNase free
Packaging Poly Bottle
Molecular Formula C3H5NO
MDL Number MFCD00008032
UN Number 2074
InChI Key HRPVXLWXLXDGHG-UHFFFAOYSA-N
SMILES NC(=O)C=C
IUPAC Name prop-2-enamide
Molecular Weight (g/mol) 71.08
PubChem CID 6579
ChEBI CHEBI:28619
Grade Electrophoresis
Chemical Name or Material Acrylamide: Bis-Acrylamide 19:1
Vedi altri risultati Mostra meno risultati
compliance-icons compliance-icons
Identificativo prodotto
  • Acrylamide/Bis-Acrylamide Mixture (19:1)
Avvertenza
  • Danger
Categoria di pericolo
  • Acute toxicity Category 3
  • Acute toxicity Category 4
  • Carcinogenicity Category 1B
  • Serious eye damage/eye irritation Category 2
  • Germ cell mutagenicity Category 1B
  • Reproductive toxicity Category 2
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 1
Indicazioni di pericolo
  • H317-May cause an allergic skin reaction.
  • H301-Toxic if swallowed.
  • H372-Causes damage to organs.
  • H340-May cause genetic defects.
  • H350-May cause cancer.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H361f-Suspected of damaging fertility.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
Consigli di prudenza
  • P201-Obtain special instructions before use.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.