missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Acrylamide/Bisacrylamide Solution (40%, 37.5:1)

Product Code. 15433569
Change view
Click to view available options
Quantity:
500 mL
1 L
Unit Size:
1 L
500mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15433569 500 mL 500mL
15443569 1 L 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.
Product Code. 15433569 Supplier Thermo Scientific Chemicals Supplier No. J60868.AP
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.

Acrylamide/Bisacrylamide Solution (40%, 37.5:1), also known as bioAcryl-P (40%, 37.5:1), is a premixed solution containing acrylamide and N,N'-methylenebisacrylamide (in a ratio of 37.5:1, respectively) used for preparing protein DNA gels.

Acrylamide/Bisacrylamide premixed solution is used for preparing protein DNA gels. It is used as a DNA sequencing gels, Protein separation. They are also used for preparing gels using stock solutions with the most common cross-link ratios.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

• Acrylamide/Bisacrylamide solution is used to to separate proteins
• The ratio of bisacrylamide to acrylamide modulates the porosity of the polyacrylamide gel

Application

• Acrylamide/Bisacrylamide Solution is used as a DNA sequencing gel promoting protein separation in the electrophoretic gels
• This solution is also applied for preparing gels using stock solutions with the most common cross-link ratios
• It has also been used as a precursor for polyacrylamide lid fabrication by photopatterning methods

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Acrylamide/Bisacrylamide 37.5:1
Physical Form Liquid
Concentration 40% soln
Color Colorless
Quantity 500 mL
MDL Number MFCD00080848
UN Number UN3426
Solubility Information It is soluble in water.
compliance-icons compliance-icons
Product Identifier
  • Acrylamide/Bisacrylamide Solution (40%, 37.5:1)
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Carcinogenicity Category 1B
  • Serious eye damage/eye irritation Category 2
  • Germ cell mutagenicity Category 1B
  • Reproductive toxicity Category 2
  • Skin corrosion/irritation Category 2
  • Skin sensitiser Category 1
  • Specific target organ toxicity Category 1
Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H317-May cause an allergic skin reaction.
  • H319-Causes serious eye irritation.
  • H340-May cause genetic defects.
  • H350-May cause cancer.
  • H361f-Suspected of damaging fertility.
  • H372-Causes damage to organs.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: Acrylamide, Methylene diacrylamide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.