missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Acrylamide / N,N'-Methylenebisacrylamide 29:1, for biochemistry, 40% mix solution in water

Product Code. 10245481 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 mL
500 mL
Unit Size:
100mL
500mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10245481 100 mL 100mL
10152961 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.
Product Code. 10245481 Supplier Thermo Scientific Chemicals Supplier No. 330211000
 Restricted Item
Estimated Shipment: 22-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.

Acrylamide / N,N'-Methylenebisacrylamide, CAS # 79-06-1, 7732-18-5, 110-26-9, 79-06-1, is a water soluble compound that cross-links to produce polyacrylamide gels that are widely used in chromatography and the electrophoretic separation of nucleic acids and proteins. | CAS: 79-06-1 | C3H5NO | 71.08 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

  • Acrylamide / N,N'-Methylenebisacrylamide is a water-soluble, odorless, white crystalline powder
  • This compound polymerizes in the presence of free radicals to generate gels whose pore-size may be regulated by manipulating the relative concentrations of this mixture of compounds

Applications

  • Polyacrylamide gels are widely used in biochemistry, forensic chemistry, genetics, molecular biology, and biotechnology to separate proteins or nucleic acids, according to their electrophoretic mobility
  • Because of their higher resolution compared to other gels, polyacrylamide gels are also used in the analysis of smaller RNA species from mixed samples
TRUSTED_SUSTAINABILITY

Specifications

Absorbance (1% in water), (1% in water) at 0nm, 0.1 max. at 290nm
Chemical Name or Material Acrylamide / N,N′-Methylenebisacrylamide 29:1
Physical Form Liquid
Acrylic Acid 0.001% max.
CAS 79-06-1, 7732-18-5, 110-26-9, 79-06-1
CAS Min % 59.0
CAS Max % 61.0
Conductivity 3μS/cm Max.
Color Colorless
Quantity 100 mL
Packaging Glass Bottle
Molecular Formula C3H5NO
MDL Number MFCD00008032
InChI Key HRPVXLWXLXDGHG-UHFFFAOYSA-N
SMILES NC(=O)C=C
IUPAC Name prop-2-enamide
Molecular Weight (g/mol) 71.08
PubChem CID 6579
ChEBI CHEBI:28619
Grade Biochemical
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.