missing translation for 'onlineSavingsMsg'
Learn More

Acrylic Acid (stabilized with MEHQ), TRC

Product Code. 30010485
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30010485 5 g 5g
30006493 100 g 100g
30009204 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30010485 Supplier Toronto Research Chemicals Supplier No. TRCA1913505G

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 79-10-7
Molecular Formula C3H4O2
Molecular Weight (g/mol) 72.06
Synonym 2-Propenoic Acid, Acroleic Acid, Ethylenecarboxylic Acid, NSC 4765, Propenoic Acid, Vinylformic Acid;
IUPAC Name prop-2-enoic acid
SMILES OC(C=C)=O

Specifications

Quantity 5 g
InChI Formula InChI=1S/C3H4O2/c1-2-3(4)5/h2H,1H2,(H,4,5)
Formula Weight 72.0211
Percent Purity >95%
Chemical Name or Material Acrylic Acid
Grade GC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.